EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Direct chloride measurement of [4-36Cl]chlorobiphenyl and [4,4'-36Cl]dichlorobiphenyl dechlorination in the rat.
PMID 6436465
Abstract
A reverse-phase HPLC system was used for the determination of inorganic chloride liberated in vivo from two biphenyl compounds in the rat. Oral administration of [4-36Cl]chlorobiphenyl resulted in a total yield of 27.6% of the original dose excreted over 10 d in the urine and included 1.7% of the dose as inorganic chloride. For [4,4'-36Cl]dichlorobiphenyl, the radioactivity of the original dose in the urine was 39.8% after 10 d, which included 11.5% of the dose as inorganic chloride. These results appear to represent the first direct determination of dechlorination by measurement of the inorganic chloride and suggest that biodechlorination plays a greater role in the metabolism of these compounds than previously expected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48260-U 4,4′-Dichlorobiphenyl solution, certified reference material