EMAIL THIS PAGE TO A FRIEND
Postepy higieny i medycyny doswiadczalnej
[Enzymoimmunologic method of determining biologically active substances and its use].
PMID 6443022
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104590
N,N′-(o-Phenylene)dimaleimide, 99%
C14H8N2O4