EMAIL THIS PAGE TO A FRIEND
Biochemistry
Effect of hydrophobic carboxyl reagents on the proton flux through coupling factor CF0 in thylakoid membrane.
PMID 6446930
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178241
Isobutyl 1,2-dihydro-2-isobutoxy-1-quinolinecarboxylate, 99%
C18H25NO3