EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
But-2-yne-1,4 diol, primary gloss improver and contact sensitizer in a nickel plating bath.
PMID 6447036
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B103209
2-Butyne-1,4-diol, 99%
C4H6O2