EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
New metabolites in isovaleric acidemia.
PMID 6452974
Abstract
Two metabolites, 4-hydroxyisovaleric acid and mesaconic acid, have been identified and quantified in the urine of a patient with isovaleric acidemia. These compounds do not appear to have been reported previously as being components of human metabolism. In addition, large quantities of 3-methylbutyrolactone, the lactone of 4-hydroxyisovaleric acid, were observed in the volatile profile obtained by headspace chromatography. The demonstration of 4-hydroxyisovaleric acid supports the contention that urinary methylsuccinic acid seen in patients with isovaleric acidemia has arisen by omega-oxidation of isovaleric acid. The identification of mesaconic acid may indicate that the methylsuccinic acid formed in these patients is subject to further metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131040
Mesaconic acid, 99%
C5H6O4