EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Labeling of thiols involved in the activity of complex V of the mitochondrial oxidative phosphorylation system.
PMID 6453870
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
373605
6,6′-Dithiodinicotinic acid, technical grade, 85%
C12H8N2O4S2