EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A novel enzyme catalyzes the synthesis of activation factor from ATP and D-fructose-6-P.
PMID 6454686
Abstract
We have recently discovered an activator for phosphofructokinase termed "activation factor" (Furuya, E., and Uyeda, K. (1980) Proc. Natl. Acad. Sci. U. S. A. 77, 5861-5864). In this paper, we investigated the enzyme responsible for its synthesis. We have found an enzyme from rat liver which catalyzes the formation of activation factor from fructose-6-P and ATP-Mg and it has been identified as fructose-2,6-P2. Fructose-1,6-P2, fructose-1-P, or fructose does not serve as a substrate. This enzyme has been partially purified and shown to be different from phosphofructokinase. Several lines of evidence indicate that the in vitro synthetic product is identical with chemically synthesized fructose-2,6-P2: (a) it is active in our assay for activation factor which is based on counteraction of ATP inhibition of phosphofructokinase; (b) it is acid labile as is fructose-2,6-P2; and (c) it shows the same mobility as synthetic fructose-2,6-P2 upon paper chromatography and the acid hydrolysis product has been identified as fructose-6-P. Thus, this new enzyme catalyzes the synthesis of the activation factor from fructose-6-P and ATP-Mg.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47748
D-(−)-Fructose, tested according to Ph.Eur.
C6H12O6