EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Occupational vitiligo: clinical and epidemiological aspects.
PMID 6455147
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
153842
4-tert-Amylphenol, 99%
C11H16O