EMAIL THIS PAGE TO A FRIEND
Thrombosis research
Relationships between the chemical constitution of carbamoylpiperidines and related compounds, and their inhibition of ADP-induced human blood platelet aggregation.
PMID 6458118
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8105
Nipecotamide, 95%
C6H12N2O