EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Occupational contact sensitization to 2,6-dichloropurine.
PMID 6461481
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36390
2,6-Dichloropurine, purum, ≥97.0% (AT)
C5H2Cl2N4
D73103
2,6-Dichloropurine, 97%
C5H2Cl2N4