EMAIL THIS PAGE TO A FRIEND
Pharmacological research communications
Beta-phenyl cysteine: a leucine aminopeptidase inhibitor.
PMID 6463094
Abstract
Beta-phenyl cysteine: a leucine aminopeptidase inhibitor. The kinetic parameters of the inhibition of the leucine aminopeptidase by beta-phenyl cysteine are reported in this communication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9125
L-Leucine-p-nitroanilide, leucine aminopeptidase substrate
C12H17N3O3
530190
S-Phenyl-L-cysteine, 97%
C9H11NO2S