EMAIL THIS PAGE TO A FRIEND
Nippon Ganka Gakkai zasshi
[Antiviral effects of 9-beta-D-arabinofuranosyl-2,6-diaminopurine against experimental herpetic keratitis].
PMID 6464878
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
766755
2,6-Diaminopurine-9-arabinoside, 97%
C10H14N6O4