EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Method of determining o-tolylbiguanide in water].
PMID 6468951
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
424668
1-(o-Tolyl)biguanide, 98%
C9H13N5