EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Methods of identifying and determining plantvax and vitavax in water].
PMID 6468988
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36185
Oxycarboxine, PESTANAL®, analytical standard
C12H13NO4S