EMAIL THIS PAGE TO A FRIEND
World review of nutrition and dietetics
Influence of dietary purines on the metabolism of allopurinol in man.
PMID 6475097
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2026
Allopurinol riboside
C10H12N4O5