EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[Allergy to dyes in stockings].
PMID 6479046
Abstract
Skin allergies caused by the wearing of stockings and hose have received little attention. Findings in patients of an allergy department, enquiries at stocking counters of stores and recent publications indicate, however, that probably many more persons have an allergy to stocking dyes than is generally thought. Skin tests with isolated stocking dyes indicate that azo dye dispersion yellow 3, dispersion orange 3 and dispersion red 1 are the most important contact allergens. They were demonstrated in 18-21 of the 23 hose examined. In textile materials, azo dye dispersion blue 124 is predominant among allergens. Cross-reactions may occur to other dispersion azo dyes, used in cosmetics, textiles, toiletries and hygenic articles, permitted food additives and hair dyes. It is suggested that in persons who have dye allergy or intolerance, decolouration followed by colouring with natural colours be undertaken.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215686
Disperse Yellow 3, Dye content 30 %
C15H15N3O2
11344
Disperse Yellow 3, analytical standard
C15H15N3O2