EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Influence of five phenylurea herbicides on the diatom Hantzschia in a sandy loam soil.
PMID 6487837
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36174
Monuron, PESTANAL®, analytical standard
C9H11ClN2O