EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Application of molecular-mechanics calculations to D-glucal and its acetate; a comparison of X-ray and n.m.r. results.
PMID 6488200
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
464058
D-Glucal, 96%
C6H10O4
T44407
Tri-O-acetyl-D-glucal, 98%
C12H16O7