EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergy to 2 new corticoid molecules.
PMID 6488784
Abstract
6 cases of hypersensitivity to Tixocortol pivalate and 3 to Amcinonide are reported. The 9 female patients all presented with features of contact dermatitis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2428
Amcinonide, analytical standard
C28H35FO7