EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
Solubility of cholesterol in bile salt-synthetic detergent mixed micelles & that of cholesteryl benzoate in synthetic detergent mixed micelles.
PMID 6490055
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C75802
Cholesteryl benzoate, 98%
C34H50O2