EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Acetylation of sulphamethoxazole by the snail Cepaea hortensis.
PMID 6492251
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000412
Sulfamethoxazole impurity A, European Pharmacopoeia (EP) Reference Standard
C12H13N3O4S