EMAIL THIS PAGE TO A FRIEND
Kaku igaku. The Japanese journal of nuclear medicine
[Development of positron tracer for in vivo estimation of brain MAO-B activity: theoretical consideration of metabolic-trapping tracers and evaluation of 11C-N,N dimethylphenylethylamine].
PMID 6492473
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
523801
N,N-Dimethylphenethylamine, 98%
C10H15N