EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Dehalogenation reactions in fast atom bombardment mass spectrometry.
PMID 6496958
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852597
5-Iodouridine, 95%
C9H11IN2O6