EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Review of glycol ether and glycol ether ester solvents used in the coating industry.
PMID 6499793
Abstract
Ethylene oxide-based glycol ether and glycol ether ester solvents have been used in the coatings industry for the past fifty years. Because of their excellent performance properties (evaporation rate, blush resistance, flow-out and leveling properties, solubility for coating resins, solvent activity, mild odor, good coupling ability, good solvent release) a complete line of ethylene oxide-based solvents of various molecular weights has been developed. These glycol ether and glycol ether ester solvents have better solvent activity for coating resin than ester or ketone solvents in their evaporation rate range. The gloss, flow and leveling, and general performance properties of many coating systems are dependent on the use of these products in the coating formula. Because of the concern about the toxicity of certain ethylene oxide-based solvents, other products are being evaluated as replacements in coating formulas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W509043
Di(ethylene glycol) ethyl ether, ≥99%
C6H14O3
537551
Ethylene glycol butyl ether, ≥99%
C6H14O2