EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Analysis of flumethasone pivalate formulations by high-performance liquid chromatography.
PMID 6501501
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0891
Flumethasone pivalate
C27H36F2O6