EMAIL THIS PAGE TO A FRIEND
Hepato-gastroenterology
Severe liver injuries due to sulfamethoxazole-trimethoprim and sulfamethoxydiazine.
PMID 6510880
Abstract
Two cases of severe sulfonamide liver injury are described. In one patient, fulminant liver failure developed after two days of taking sulfamethoxazole-trimethoprim. The patient was treated with resin hemoperfusion and recovered completely. Another patient became jaundiced after using sulfamethoxydiazine for ten days. Very severe intrahepatic cholestasis developed and cleared up only after high-dose prednison treatment. Marked hyperbilirubinemia persisted for four months and aminotransferase activity had not normalized ten months after the onset of the disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0383
Sulfameter
C11H12N4O3S
46834
Sulfameter, VETRANAL®, analytical standard
C11H12N4O3S