EMAIL THIS PAGE TO A FRIEND
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan
[Studies on the synthesis of anti-inflammatory activity of 2,6-di-tert-butylphenols with a heterocyclic group at the 4-position. IV. Photo-induced and base-catalyzed oxygenation of 4-(3,5-di-tert-butyl-4-hydroxyphenyl)-2-oxo-4-imidazolines].
PMID 6520717
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D48400
2,6-Di-tert-butylphenol, 99%
C14H22O