EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series
Studies on oxidative modifications of nucleic acid pyrimidine bases.
PMID 6522278
Abstract
Oxidation of diacetylthymidine (4), triacetyluridine (5), and 5-fluoro-1,3-dimethyluracil (3) with m-chloroperbenzoic acid and cross-linkage of the bromohydrin (6) with the benzylamine and glycine ethyl ester are described. A Plausible mechanism for formation of oxidation products is also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
406864
5-Fluoro-1,3-dimethyluracil, 99%
C6H7FN2O2