EMAIL THIS PAGE TO A FRIEND
Steroids
Synthesis of a precursor for the preparation of 9 alpha,11 alpha-tritiated 5 alpha-androstane-3 alpha,17 beta-diol 17-glucuronide.
PMID 6523548
Abstract
Starting from 11 beta-hydroxytestosterone, we achieved the synthesis of a strategic precursor, C-9 (11) unsaturated 5 alpha-androstane-3 alpha, 17 beta-diol 17-glucuronide (9a), for the preparation of 9 alpha,11 alpha-tritiated 5 alpha-androstane-3 alpha, 17 beta-diol 17-glucuronide. We optimized the reaction conditions for catalytic reduction employing hydrogen and subsequent base hydrolysis followed by purification on Amberlite XAD-2 resin to obtain the saturated 5 alpha-androstane-3 alpha, 17 beta-diol 17-glucuronide.