EMAIL THIS PAGE TO A FRIEND
The Analyst
High-sensitivity extraction-spectrophotometric determination of iron with 3-(2-pyridyl)-5,6-diphenyl-1,2,4-triazine and tetrabromophenolphthalein ethyl ester.
PMID 6529001