EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Hygienic regulation of the monurone content of soil].
PMID 6530160
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36174
Monuron, PESTANAL®, analytical standard
C9H11ClN2O