EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Effect of seneciphylline and senecionine on hepatic drug metabolizing enzymes in rats.
PMID 6533413
Abstract
The effect of oral administration of the pyrrolizidine alkaloids, seneciphylline and senecionine, from Senecio vulgaris (Compositae) on activities of hepatic epoxide hydrase, glutathione-S-transferase, aminopyrine-N-demethylase and arylhydrocarbon hydroxylase (AHH) was investigated in microsomes of young male albino rats. Seneciphylline significantly increased the activities of epoxide hydrase and glutathione-S-transferase but caused reduction of cytochrome P-450 and related monooxygenase activities. Senecionine failed to stimulate epoxide hydrase while it diminished the activities of glutathione-S-transferase, aminopyrine demethylase and AHH. Seneciphylline and senecionine could not produce any prominent in vitro effect on the hepatic drug metabolizing enzymes under study, except slight stimulation of epoxide hydrase activity by both the alkaloids and slight reduction of aminopyrine demethylase activity by senecionine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73913
Seneciphylline, analytical standard
C18H23NO5