EMAIL THIS PAGE TO A FRIEND
Mutation research
Isomeric mononitrobenzo[a]pyrenes: synthesis, identification and mutagenic activities.
PMID 6540365
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR311
6-Nitrobenzo[a]pyrene, BCR® certified Reference Material
C20H11NO2