EMAIL THIS PAGE TO A FRIEND
Annals of internal medicine
Vitamin D conversion by sarcoid lymph node homogenate.
PMID 6546329
Abstract
Recent studies suggest that hypercalcemia of sarcoidosis is due to high blood concentrations of calcitriol and that this compound may be synthesized at an extra-renal site. We report that sarcoid lymph node homogenate metabolized calcifediol to a substance indistinguishable from calcitriol, whereas six nonsarcoid lymph nodes failed to produce this compound.