EMAIL THIS PAGE TO A FRIEND
The International journal of applied radiation and isotopes
Coupling antibody with DTPA--an alternative to the cyclic anhydride.
PMID 6547405
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284025
Diethylenetriaminepentaacetic dianhydride, 98%
C14H19N3O8