EMAIL THIS PAGE TO A FRIEND
International journal of nuclear medicine and biology
Tumor uptake of radiolabelled pyrimidine bases and pyrimidine nucleosides in animal models--V. 6-[36Cl]chlorouracil, 6-[82Br]bromouracil and 6-[123I]iodouracil.
PMID 6549392
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9525
4-Chlorouracil, ≥99%
C4H3ClN2O2