EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A simple method for stabilizing protein-sulfhydryl groups during SDS-gel electrophoresis.
PMID 655420
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43794
1,4-Dithioerythritol, BioUltra, ≥99.0% (RT)
C4H10O2S2