EMAIL THIS PAGE TO A FRIEND
Schweizerische medizinische Wochenschrift
Protective effect of specific elastase inhibitor in experimental pulmonary emphysema.
PMID 6565360
Abstract
The inhibitory effect of synthetic elastase inhibitor was studied in experimental emphysema. The development of emphysema was totally prevented by this substance. According to the authors' opinion the administration of synthetic elastase inhibitors can be considered as a new possibility in the treatment of pulmonary emphysema.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0398
N-(Methoxysuccinyl)-Ala-Ala-Pro-Val-chloromethyl ketone, elastase inhibitor
C22H35ClN4O7