EMAIL THIS PAGE TO A FRIEND
Dakar medical
[Evaluation of bird poison residues in "millet-eaters" (Quelea quelea L.). Toxicologic consequences arising from the consumption of poisoned birds by domestic animals and man. 2. Cyanophos].
PMID 6571258
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46279
Cyanophos, PESTANAL®, analytical standard
C9H10NO3PS