EMAIL THIS PAGE TO A FRIEND
Caries research
Effect of surface-active agents on fluoride enamel interactions. II.
PMID 6575867
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4266
3-(Decyldimethylammonio)­propane­sulfonate inner salt, zwitterionic detergent
C15H33NO3S