EMAIL THIS PAGE TO A FRIEND
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement
Fluoroacetamide poisoning in man: the role of ionized calcium.
PMID 6578726
Abstract
Two cases are reported of severe acute fluoroacetamide poisoning in man, with successful treatment of the life-threatening cardiac arrhythmias by the administration of calcium chloride. The arrhythmias were preceded by prolongation of the QT interval in the ECG. Calcium chloride therapy restored to normal the markedly prolonged QT interval.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
128341
Fluoroacetamide
C2H4FNO