EMAIL THIS PAGE TO A FRIEND
Deutsche zahnarztliche Zeitschrift
[Efficiency of fluoride dosage forms: solutions and gels].
PMID 6580164
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205567
Zinc fluoride, powder, 99%
F2Zn