EMAIL THIS PAGE TO A FRIEND
Shigaku = Odontology; journal of Nihon Dental College
[Basic studies on the clinical application of cresatin].
PMID 6585761
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
531677
m-Tolyl acetate, 97%
C9H10O2