EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans
9-Nitroanthracene.
PMID 6590451
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N10209
9-Nitroanthracene, 93%
C14H9NO2
BCR308
9-Nitroanthracene, BCR® certified Reference Material
C14H9NO2