EMAIL THIS PAGE TO A FRIEND
Shikai tenbo = Dental outlook
[Guaiazulene mouthwash for diseases of the oral mucosa].
PMID 6591496
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G11004
Guaiazulene, 99%
C15H18