EMAIL THIS PAGE TO A FRIEND
Caries research
Effects of chlorhexidine, iodine, and 5,7-dichloro-8-hydroxyquinoline on the bacterial composition of rat plaque in vivo.
PMID 6592046
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D64600
5,7-Dichloro-8-quinolinol, 99%
C9H5Cl2NO