EMAIL THIS PAGE TO A FRIEND
Proceedings of the Western Pharmacology Society
Design of ionophore hapten conjugates for electroimmunoassay.
PMID 6593738
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158399
Dibenzo-18-crown-6, 98%
C20H24O6
49902
Tin(II) ionophore I, Selectophore
C20H24O6