EMAIL THIS PAGE TO A FRIEND
Iyo Kizai Kenkyujo hokoku. Reports of the Institute for Medical and Dental Engineering, Tokyo Medical and Dental University
[Properties of 2,2,2-Trifluoroethyl methacrylate copolymers].
PMID 6598491
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
373761
2,2,2-Trifluoroethyl methacrylate, contains 50-200 ppm MEHQ as inhibitor, 99%
C6H7F3O2