EMAIL THIS PAGE TO A FRIEND
Basic life sciences
Suicide enzyme inactivators.
PMID 6602608
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I12808
Isatoic anhydride, 96%
C8H5NO3