EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Assessment of the chloroethyl analog of 3-PPP (N-n-propyl-3-(3-hydroxyphenyl)-piperidine), 3-PPP-C1 as an irreversible ligand for central dopaminergic recognition sites.
PMID 6617745
Abstract
NCA, the chloro analog of the potent dopamine agonist NPA is an irreversible ligand at dopamine receptors in mammalian brain. The chloroethyl analog of the recently described putative dopamine autoreceptor agonist 3-PPP, 3-PPP-C1, was evaluated for its potential use as an irreversible autoreceptor ligand. N-Chloroethylation of 3-PPP reduced the intrinsic affinity of the agonist seven-fold and, consequently, in contrast to NCA, it was found that 3-PPP-C1 was not a good irreversible ligand at dopamine receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D007
R(−)-Chloroethylnorapomorphine hydrochloride, analytical standard
C18H18ClNO2 · HCl